Learn More
Medchemexpress LLC HY-13016 50mg Medchemexpress, Cabozantinib CAS:849217-68-1 Purity:>98%
SDP
Catalog No. 502028942
Encompass_Preferred
Medchemexpress, HY-13016 50mg Cabozantinib CAS:849217-68-1 Cabozantinib is a potent multiple receptor tyrosine kinases (RTKs) inhibitor that inhibits VEGFR2, c-Met, Kit, Axl and Flt3 with IC50s of 0.035, 1.3, 4.6, 7 and 11.3 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8942 Supplier Medchemexpress LLC Supplier No. HY1301650MG
Medchemexpress, HY-13016 50mg Cabozantinib CAS:849217-68-1 Cabozantinib is a potent multiple receptor tyrosine kinases (RTKs) inhibitor that inhibits VEGFR2, c-Met, Kit, Axl and Flt3 with IC50s of 0.035, 1.3, 4.6, 7 and 11.3 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.