Learn More
Medchemexpress LLC Cabozantinib | 849217-68-1 | >98% | 500 MG
SDP
Catalog No. 5000340995
Encompass_Preferred
Cabozantinib is a potent and orally active inhibitor of VEGFR2 and MET. It displays strong inhibition of KIT, RET, AXL, TIE2, and FLT3. Cabozantinib shows antiangiogenic activity, disrupts tumor vasculature, and promotes tumor and endothelial cell apoptosis.
  • Potent and orally active inhibitor of VEGFR2 and MET
  • Displays strong inhibition of KIT, RET, AXL, TIE2, and FLT3
  • Shows antiangiogenic activity
  • Disrupts tumor vasculature
  • Promotes tumor and endothelial cell apoptosis
Catalog No. 50-003-40995 Supplier Medchemexpress LLC Supplier No. HY13016500MG
Cabozantinib is a potent and orally active inhibitor of VEGFR2 and MET. It displays strong inhibition of KIT, RET, AXL, TIE2, and FLT3. Cabozantinib shows antiangiogenic activity, disrupts tumor vasculature, and promotes tumor and endothelial cell apoptosis.
  • Potent and orally active inhibitor of VEGFR2 and MET
  • Displays strong inhibition of KIT, RET, AXL, TIE2, and FLT3
  • Shows antiangiogenic activity
  • Disrupts tumor vasculature
  • Promotes tumor and endothelial cell apoptosis