Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cabozantinib-d4 | 1802168-53-1 | 99.0% | 505.53 g/mol | C28H20D4FN3O5 | 5 MG
SDP

Catalog No. 5000263160
Encompass_Preferred

Cabozantinib-d4 is the deuterium-labeled (d4) form of cabozantinib used as a stable-isotope internal standard for quantitative LC-MS and bioanalytical assays. It is supplied as a high-purity solid suitable for method development, calibration, and pharmacokinetic studies.

  • Deuterium-labeled internal standard for LC-MS analysis.
  • High purity suitable for quantitative assays.
  • Supplied in small-mass quantities for analytical workflows.
  • Molecular formula C28H20D4FN3O5 and molecular weight 505.53 g/mol.
  • Intended for research use and analytical method development.

Catalog No. 50-002-63160 Supplier Medchemexpress LLC Supplier No. HY13016S15MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cabozantinib-d4 is the deuterium-labeled (d4) form of cabozantinib used as a stable-isotope internal standard for quantitative LC-MS and bioanalytical assays. It is supplied as a high-purity solid suitable for method development, calibration, and pharmacokinetic studies.

  • Deuterium-labeled internal standard for LC-MS analysis.
  • High purity suitable for quantitative assays.
  • Supplied in small-mass quantities for analytical workflows.
  • Molecular formula C28H20D4FN3O5 and molecular weight 505.53 g/mol.
  • Intended for research use and analytical method development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.