Learn More
Medchemexpress LLC Cabozantinib-d6 | 1802168-46-2 | 98.1% | 25 MG
SDP
Catalog No. 5000338796
Encompass_Preferred
Cabozantinib-d6 is a deuterium-labeled version of Cabozantinib, a potent inhibitor of multiple receptor tyrosine kinases (RTKs), including VEGFR2, c-Met, Kit, Axl, and Flt3. It is intended for research use only and not sold to patients. This compound can be used as a tracer or as an internal standard for quantitative analysis.
  • Used as a tracer or internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Potent inhibitor of multiple receptor tyrosine kinases (RTKs).
  • Inhibits VEGFR2, c-Met, Kit, Axl, and Flt3.
  • Deuterium-labeled for use as tracers in drug development.
  • May impact pharmacokinetic and metabolic profiles of drugs.
  • For research use only.
Catalog No. 50-003-38796 Supplier Medchemexpress LLC Supplier No. 25MGHY13016S
Add to Cart
Add to Cart
Cabozantinib-d6 is a deuterium-labeled version of Cabozantinib, a potent inhibitor of multiple receptor tyrosine kinases (RTKs), including VEGFR2, c-Met, Kit, Axl, and Flt3. It is intended for research use only and not sold to patients. This compound can be used as a tracer or as an internal standard for quantitative analysis.
  • Used as a tracer or internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Potent inhibitor of multiple receptor tyrosine kinases (RTKs).
  • Inhibits VEGFR2, c-Met, Kit, Axl, and Flt3.
  • Deuterium-labeled for use as tracers in drug development.
  • May impact pharmacokinetic and metabolic profiles of drugs.
  • For research use only.