Learn More
Medchemexpress LLC Cabozantinib-d6 | 1802168-46-2 | 98.14% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY13016S100MG
Encompass_Preferred
Cabozantinib-d6 is the deuterium labeled Cabozantinib. Cabozantinib is a potent multiple receptor tyrosine kinases (RTKs) inhibitor that inhibits VEGFR2, c-Met, Kit, Axl and Flt3 with IC50s of 0.035, 1.3, 4.6, 7 and 11.3 nM, respectively.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-39044