Learn More
Medchemexpress LLC Cabozantinib-d6 | 1802168-46-2 | 98.14% | 507.54 | 5 MG
SDP
Catalog No. 5000364232
Encompass_Preferred
Cabozantinib-d6 is the deuterium labeled form of Cabozantinib, a potent inhibitor of multiple receptor tyrosine kinases (RTKs). It is suitable for use as a tracer or as an internal standard for quantitative analysis in various analytical methods.
  • Inhibits VEGFR2, c-Met, Kit, Axl, and Flt3.
  • Used as a tracer for quantitation during the drug development process.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs.
  • For research use only and not for medical applications.
  • Store at 4°C, protected from light.
Catalog No. 50-003-64232 Supplier Medchemexpress LLC Supplier No. 5MGHY13016S
Cabozantinib-d6 is the deuterium labeled form of Cabozantinib, a potent inhibitor of multiple receptor tyrosine kinases (RTKs). It is suitable for use as a tracer or as an internal standard for quantitative analysis in various analytical methods.
  • Inhibits VEGFR2, c-Met, Kit, Axl, and Flt3.
  • Used as a tracer for quantitation during the drug development process.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs.
  • For research use only and not for medical applications.
  • Store at 4°C, protected from light.