Learn More
Apexbio Technology LLC Cabozantinib malate (XL184), 5mg. Cas: 1140909-48-3 MFCD: MFCD20923480. MET andVEGF receptor2 inhibitor
SDP
Catalog No. 501014580
Encompass_Preferred
Cabozantinib malate is a potent inhibitor of MET andVEGF receptor2 with IC50 values of 1.3nM and 0.035nM. 1140909-48-3. MFCD20923480
Catalog No. 50-101-4580 Supplier Apexbio Technology LLC Supplier No. B14015
Cabozantinib malate is a potent inhibitor of MET andVEGF receptor2 with IC50 values of 1.3nM and 0.035nM. 1140909-48-3. MFCD20923480