Learn More
Apexbio Technology LLC Cabozantinib (XL184, BMS-907351), 5mg. Cas: 849217-68-1 MFCD: MFCD20926324. VEGFR2/Met/Ret/Kit/FLT//AXL inhibitor
SDP
Supplier:  Apexbio Technology LLC A29775
Encompass_Preferred
Cabozantinib (also called XL184, BMS-907351 Cometriq [1]), is an inhibitor of multiple receptor tyrosine kinases (RTKs), including vascular endothelial growth factor receptor 2 (VEGFR2), hepatocyte growth factor receptor (MET), and rearranged during transfection (RET) [2] [3]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4769