Learn More
Santa Cruz Biotechnology CABS BUFFER
Catalog No. NC1107635
Encompass
Application: A buffering agent useful in the pH range of 10.0 - 11.4 ; CAS Number: 161308-34-5 ;Purity: ≥98% ;Molecular Weight: 235.34 ; Unit 25g
Catalog No. NC1107635 Supplier Santa Cruz Biotechnology Supplier No. SC285279
Application: A buffering agent useful in the pH range of 10.0 - 11.4 ; CAS Number: 161308-34-5 ;Purity: ≥98% ;Molecular Weight: 235.34 ; Unit 25g