Learn More
Aviva Systems Biology Corp CABYR Antibody - middle region (ARP53673_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53673_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CABYR. It was validated on Western Blot. . Alternate names: CBP86, CT88, FSP-2, FSP2, MGC9117, CABYRa, CABYRc, CABYRe, CABYRc/dApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-869