Learn More
New England Biolabs, Inc. Cac8I – 500 units
SDP
Catalog No. 50812279
Encompass
A restriction endonuclease that recognizes the sequence GCN_^NGC.
Catalog No. 50-812-279 Supplier New England Biolabs, Inc. Supplier No. R0579L
A restriction endonuclease that recognizes the sequence GCN_^NGC.