Learn More
Promocell GmbH PromoCell CaCl 2-Solution 2 ml, 0.5 M
Catalog No. 50306304
Encompass_Preferred
Some types of primary human cells, especially Normal Human Epidermal Keratinocytes (NHEK), respond sensitively to the calcium concentration in the culture medium. Calcium Chloride solution allows the adjustment of the optimal amount of calcium required by the cells. PromoCell Calcium Chloride (CaCl~2~) Solution is prepared as a solution from highly pure tissue culture tested Calcium Chloride and cell culture-grade water. It is a clear colorless and sterile solution.The concentration of CaCl~2~ is 0.5 M.
Catalog No. 50-306-304 Supplier Promocell GmbH Supplier No. C34006
Some types of primary human cells, especially Normal Human Epidermal Keratinocytes (NHEK), respond sensitively to the calcium concentration in the culture medium. Calcium Chloride solution allows the adjustment of the optimal amount of calcium required by the cells. PromoCell Calcium Chloride (CaCl~2~) Solution is prepared as a solution from highly pure tissue culture tested Calcium Chloride and cell culture-grade water. It is a clear colorless and sterile solution.The concentration of CaCl~2~ is 0.5 M.