Learn More
CELLINK LLC CaCl2 Crosslinking Agent - Sterile Bottle 1x45 ml
SDP
Catalog No. NC3733364
Encompass
Crosslinking Agent contains 50 mM calcium chloride to ionically crosslink bioinks containing alginate.Shelf life: minimum 6 months.Storage conditions: 2-25°C, do not freeze.For research use only. Not for human use.
Catalog No. NC3733364 Supplier CELLINK LLC Supplier No. CLBICRLCCL45
Crosslinking Agent contains 50 mM calcium chloride to ionically crosslink bioinks containing alginate.Shelf life: minimum 6 months.Storage conditions: 2-25°C, do not freeze.For research use only. Not for human use.