Learn More
Aviva Systems Biology Corp CACNA1G Antibody - C - terminal region (ARP35566_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35566_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CACNA1G. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human CACNA1G. Alternate names: Ca(V)T.1, Cav3.1, MGC117234, NBR13ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55780