Learn More
Aviva Systems Biology Corp CACNA1I Antibody - middle region (ARP35319_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35319_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CACNA1I. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CACNA1I. Alternate names: Cav3.3, KIAA1120, ca(v)3.3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55696