Learn More
Aviva Systems Biology Corp CACNB1 Antibody - middle region (ARP35580_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35580_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CACNB1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CACNB1. Alternate names: CAB1, CACNLB1, CCHLB1, MGC41896ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55784