Learn More
Aviva Systems Biology Corp CACNB1 Antibody - C-terminal region (ARP36744_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36744_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CACNB1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human CACNB1. Alternate names: CAB1, CACNLB1, CCHLB1, MGC41896ApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Mouse, Rabbit, Zebrafish
Catalog No. 50-014-56263