Learn More
Aviva Systems Biology Corp CACNB2 Antibody - middle region (ARP35602_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35602_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CACNB2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CACNB2. Alternate names: CACNLB2, FLJ23743, MYSB, CAVB2ApplicationWB. Species reactivity: Human
Catalog No. 50-284-467