Learn More
Aviva Systems Biology Corp CACNB3 Antibody - C-terminal region (ARP34957_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34957_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CACNB3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human CACNB3. Alternate names: CAB3, CACNLB3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55615