Learn More
Aviva Systems Biology Corp CACNB3 Antibody - N-terminal region (ARP34958_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34958_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CACNB3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the n terminal region of human CACNB3. Alternate names: CAB3, CACNLB3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55616