Learn More
Aviva Systems Biology Corp CACNB4 Antibody - middle region (ARP36763_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36763_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CACNB4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CACNB4. Alternate names: EA5, EJM, CAB4, EIG9, EJM4, EJM6, CACNLB4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56269