Learn More
Aviva Systems Biology Corp CACNB4 Antibody - C-terminal region (ARP34959_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34959_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CACNB4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human CACNB4. Alternate names: CAB4, CACNLB4, EA5, EJM, EIG9, EJM4, EJM6ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55617