Learn More
Aviva Systems Biology Corp Cacng8 Antibody - middle region (ARP35367_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35367_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Cacng8. It was validated on Western Blot. . Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rat
Catalog No. 50-284-459