Learn More
Cayman Chemical Cacodylic Acid (sodium salt hydrate), Purity: ≥95%, CAS: 6131-99-3, 50 g
SDP
Catalog No. 502995449
Encompass_Preferred
A reagent used in electron microscopy techniques; has been used to introduce arsenic into proteins for SAD phasing in protein crystallography; has been used as an herbicide/defoliant; increases the likelihood of tumor formation in rats exposed to carcinogens at concentrations as low as 50 ppm
Catalog No. 50-299-5449 Supplier Cayman Chemical Supplier No. 2299650 G
A reagent used in electron microscopy techniques; has been used to introduce arsenic into proteins for SAD phasing in protein crystallography; has been used as an herbicide/defoliant; increases the likelihood of tumor formation in rats exposed to carcinogens at concentrations as low as 50 ppm