Learn More
Aviva Systems Biology Corp CACYBP Antibody - middle region (ARP51259_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51259_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CACYBP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CACYBP. Alternate names: GIG5, MGC87971, PNAS-107, RP1-102G20.6, S100A6BP, SIPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60528