Learn More
Selleck Chemical LLC Cadaverine dihydrochloride
SDP
Catalog No. 5000004073
Encompass_Preferred
1 5-Diaminopentane (Cadaverine) is a foul-smelling diamine formed by bacterial decarboxylation of lysine that occurs during protein hydrolysis during putrefaction of animal tissue
Catalog No. 50-000-04073 Supplier Selleck Chemical LLC Supplier No. S635725MG
1 5-Diaminopentane (Cadaverine) is a foul-smelling diamine formed by bacterial decarboxylation of lysine that occurs during protein hydrolysis during putrefaction of animal tissue