Learn More
Medchemexpress LLC HY-100436 5mg Medchemexpress, Cadazolid CAS:1025097-10-2 Purity:>98%
SDP
Catalog No. 501931833
Encompass_Preferred
Medchemexpress, HY-100436 5mg Cadazolid CAS:1025097-10-2 Cadazolid (ACT-179811) is a new oxazolidinone antibiotic with potent activity against Clostridium difficile. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1833 Supplier Medchemexpress LLC Supplier No. HY1004365MG
Medchemexpress, HY-100436 5mg Cadazolid CAS:1025097-10-2 Cadazolid (ACT-179811) is a new oxazolidinone antibiotic with potent activity against Clostridium difficile. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.