Learn More
Medicus Health Caddy and Two Wire Media Plate Holders Combo, 12.5”L x 10”W x 11”H
SDP
Catalog No. NC2163906
Encompass
∙ Culture Plate Rack Caddies are lightweight, rust free, stainless steel media plate holders with a capacity of 96 petri dishes ∙ These higher capacity racks hold up to 96 culture plates and will improve workflow and organization in the lab∙ Caddies with wire racks can be stacked on top of each other∙ Wire racks can be safely stacked with nesting tabs to maximize incubator space ∙ Racks are suitable for storing, incubating, sorting and finally autoclaving media plates ∙ Wire design lets you clearly visualize culture dishes ∙ Caddy & Wire Racks are fully autoclavable∙ By ordering this item (5310M3) you get one Caddy and two Wire Racks ∙ Overall Size: 12-1/2” L x 10” W x 11” H
This item is not returnable. View return policy
Catalog No. NC2163906 Supplier Medicus Health Supplier No. 5310M3
∙ Culture Plate Rack Caddies are lightweight, rust free, stainless steel media plate holders with a capacity of 96 petri dishes ∙ These higher capacity racks hold up to 96 culture plates and will improve workflow and organization in the lab∙ Caddies with wire racks can be stacked on top of each other∙ Wire racks can be safely stacked with nesting tabs to maximize incubator space ∙ Racks are suitable for storing, incubating, sorting and finally autoclaving media plates ∙ Wire design lets you clearly visualize culture dishes ∙ Caddy & Wire Racks are fully autoclavable∙ By ordering this item (5310M3) you get one Caddy and two Wire Racks ∙ Overall Size: 12-1/2” L x 10” W x 11” H