Learn More
AVANTOR MASTERFLEX (VWR LLC) Cadence One-way stopcocks; male luer lock x male luer lock, nickel-plated brass 41507-02
Catalog No. NC1320858
Encompass
Exceptional durability. Easy to disassemble for cleaning. Stopcock handles turn 180. Autoclavable.
Catalog No. NC1320858 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX4150702
Exceptional durability. Easy to disassemble for cleaning. Stopcock handles turn 180. Autoclavable.