Learn More
Cadence Science™ Micro-Mate Adapter
SDP
Catalog No. 1463099 Shop All Cadence Science Inc. Products
Change view
Click to view available options
Thread Size:
0.25 in. NPT
Catalog No. Thread Size
14-630-99 0.25 in. NPT
1 options
Catalog No. 14-630-99 Supplier Cadence Science™ Supplier No. 6331IND
 Restricted Item
Specifications
Certifications/Compliance ISO 594
Material Plated Brass
For Use With Male luer lock to 1/4 in. NPT
Thread Size 0.25 in. NPT
Product Type Threaded End Adapter
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov