Learn More
Diagnostic Biosystems CADHERIN 17 Cadherin 17
Catalog No. 502407391
Encompass_Preferred
Cadherin 17 is exclusively expressed in the epithelial cells of embryonic and adult small intestine, colon, liver, and pancreatic ducts. It has been reported to be frequently expressed in adenocarcinomas arising in the gastrointestinal tract and pancreas. Owing to its restricted expression in these groups of tumors, cadherin 17 has proven to be a useful immunohistochemical marker for assisting in distinguishing these neoplasms from other malignancies with which they may be confused
Catalog No. 50-240-7391 Supplier Diagnostic Biosystems Supplier No. MOB604R01
Cadherin 17 is exclusively expressed in the epithelial cells of embryonic and adult small intestine, colon, liver, and pancreatic ducts. It has been reported to be frequently expressed in adenocarcinomas arising in the gastrointestinal tract and pancreas. Owing to its restricted expression in these groups of tumors, cadherin 17 has proven to be a useful immunohistochemical marker for assisting in distinguishing these neoplasms from other malignancies with which they may be confused