Learn More
Sino Biological Recombinant Anti-Cadherin-17 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 11360R029100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Cadherin-17 / LI-cadherin / CDH17 (rh Cadherin-17 / LI-cadherin / CDH17; Catalog#11360-H08H; Q12864-1; Met1-Met787).
Catalog No. 50-228-2831