Learn More
ACROBiosystems Recombinant Protein;Human Cadherin-6 / KCAD / CDH6 Protein;HEK293;100UG;CA6,CDH6,CAD6,KCAD,K-cadherin,Cadherin-6,CDH-6
Catalog No. 501658997
Encompass_Preferred
Recombinant Protein;Human Cadherin-6 / KCAD / CDH6 Protein;100UG;Human Cadherin-6, His Tag (CA6-H5229) is expressed from human 293 cells (HEK293). It contains AA Thr 19 - Ala 615 (Accession # AAH00019).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 67.2 kDa. The protein migrates as 90-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8997 Supplier ACROBiosystems Supplier No. CA6H5229100UG
Recombinant Protein;Human Cadherin-6 / KCAD / CDH6 Protein;100UG;Human Cadherin-6, His Tag (CA6-H5229) is expressed from human 293 cells (HEK293). It contains AA Thr 19 - Ala 615 (Accession # AAH00019).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 67.2 kDa. The protein migrates as 90-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.