Learn More
Life Diagnostics Inc MOUSE CARDIAC TROPONIN-I ELISA, CTNI-1-HS
SDP
Supplier:  Life Diagnostics Inc CTNI1HS
Encompass_Preferred
ELISA kits for detection of cardiac troponin-I, myoglobin and fatty acid binding protein (FABP). Cardiac troponin-I is expressed only in cardiac muscle and therefore the best biomarker for assessment of heart damage. Myoglobin and FABP are expressed in both heart and skeletal muscle. They provide good cardiac biomarkers in the absence of skeletal muscle damage. Likewise, they serve as good skeletal muscle biomarkers in the absence of cardiac muscle damage. Advantages of myoglobin and FABP are that they are generally released at higher levels and they are also more stable than troponin-I.
Catalog No. NC9402211