Learn More
Medchemexpress LLC δ-Cadinene | 483-76-1 | 204.35 | 250 MG
SDP
Catalog No. 5000389457
Encompass_Preferred
δ-Cadinene is a sesquiterpene found in essential oils. It exhibits antiproliferative and pro-apoptotic effects on human ovarian cancer (OVCAR-3) cells, and also possesses trichomonacidal, antimicrobial, antifungal, and anticancer properties.
  • Sesquiterpene found in essential oils
  • Exhibits antiproliferative effects on human ovarian cancer (OVCAR-3) cells
  • Induces pro-apoptotic effects on human ovarian cancer (OVCAR-3) cells
  • Has trichomonacidal properties
  • Demonstrates antimicrobial properties
  • Possesses antifungal properties
  • Shows anticancer properties
  • Inhibits the growth of OVCAR-3 cells in a concentration-dependent manner
  • Induces both early and late apoptosis in a concentration-dependent manner in OVCAR-3 cells
  • Induces sub-G1-phase growth arrest in OVCAR-3 cells
Catalog No. 50-003-89457 Supplier Medchemexpress LLC Supplier No. 250MGHYN10544
δ-Cadinene is a sesquiterpene found in essential oils. It exhibits antiproliferative and pro-apoptotic effects on human ovarian cancer (OVCAR-3) cells, and also possesses trichomonacidal, antimicrobial, antifungal, and anticancer properties.
  • Sesquiterpene found in essential oils
  • Exhibits antiproliferative effects on human ovarian cancer (OVCAR-3) cells
  • Induces pro-apoptotic effects on human ovarian cancer (OVCAR-3) cells
  • Has trichomonacidal properties
  • Demonstrates antimicrobial properties
  • Possesses antifungal properties
  • Shows anticancer properties
  • Inhibits the growth of OVCAR-3 cells in a concentration-dependent manner
  • Induces both early and late apoptosis in a concentration-dependent manner in OVCAR-3 cells
  • Induces sub-G1-phase growth arrest in OVCAR-3 cells