Learn More
Description
This recombinant protein can be used for WB. For research use only.
Specifications
Specifications
| Accession Number | NP_055148.3 |
| For Use With (Application) | Western Blot |
| Gene ID (Entrez) | 23705 |
| Molecular Weight (g/mol) | 37.9 kDa |
| Quantity | 0.1 mg |
| Source | Human Cells |
| Common Name | cell adhesion molecule 1 |
| Gene Symbol | CADM1 |
| Product Type | Recombinant Protein |
| Conjugate | Unconjugated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
