Learn More
Aviva Systems Biology Corp CADM3 Antibody - middle region (ARP49641_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49641_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CADM3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human CADM3. Alternate names: BIgR, FLJ10698, IGSF4B, NECL1, Necl-1, TSLL1, synCAM3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-440