Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium AA Standard, 1000 ppm Cd in 3% HCl, Ricca Chemical
SDP

Catalog No. ACD1KH100 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
ACD1KH-100 100 mL Natural LDPE Bottle
ACD1KH-500 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ACD1KH-100 Supplier Ricca Chemical Company Supplier No. ACD1KH100
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7440-43-9 , 7647-01-0 , 7440-43-9
Molecular Formula Cd
Molecular Weight (g/mol) 112.41
MDL Number MFCD00010914
InChI Key BDOSMKKIYDKNTQ-UHFFFAOYSA-N
PubChem CID 23973
ChEBI CHEBI:22977
IUPAC Name cadmium
SMILES [Cd]

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm Cd
CAS Min % 94.0, 3.88, 0.09
CAS Max % 98.0, 4.04, 0.1
Equivalence 1 mL = 1 mg Cd
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Quantity 100 mL
Chemical Name or Material Cadmium AA Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.