Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium Acetate, Crystal, 98%, Spectrum™ Chemical
SDP

Catalog No. 18609394 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-609-394 45 kg Poly Bottle
18-609-389 25 g Poly Bottle
18-609-390 125 g Poly Bottle
18-609-392 500 g Poly Bottle
18-609-393 2.5 kg Poly Bottle
18-609-391 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-609-394 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C101545KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5743-04-4
Molecular Formula C4H10CdO6
Molecular Weight (g/mol) 266.53
InChI Key AUIZLSZEDUYGDE-UHFFFAOYSA-L
IUPAC Name cadmium(2+) diacetate dihydrate
SMILES O.O.[Cd++].CC([O-])=O.CC([O-])=O

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 45 kg
Percent Purity 0.98
Grade Reagent
Insoluble Matter 0.0001
Physical Form Crystals or Flakes
Chemical Name or Material Cadmium Acetate, Crystal

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.