Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium acetate dihydrate, 99.999% (metals basis)

Catalog No. AA1186514
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1186514 25 g
AA1186522 100 g
AA1186536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1186514 Supplier Thermo Scientific Chemicals Supplier No. 01186514
Add to Cart

CAS: 5743-04-4 | C4H10CdO6 | 266.53 g/mol

It is used in dyestuff intermediates, ceramics and producing iridescent effects on porcelains and pottery. It acts as a reagent for determination of S, Se, and Te. It is also used in cadmium electroplating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5743-04-4
Molecular Formula C4H10CdO6
Molecular Weight (g/mol) 266.53
MDL Number MFCD00012449
InChI Key AUIZLSZEDUYGDE-UHFFFAOYSA-L

Specifications

Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.999% (Metals basis)
Solubility Information Soluble in water,ethanol and methanol.
Formula Weight 266.54 (230.50 Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Density 2.01
Chemical Name or Material Cadmium acetate dihydrate
Hazard Category H302+H312+H332-H335-H350-H373
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c
DOTInformation DOT Class: 6.1, Packaging Group: III
EINECSNumber 208-853-2
RTECSNumber AF7505000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.