Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium bromide, anhydrous, 98%

Catalog No. AA3296022
Click to view available options
Quantity:
25 g
100 g

CAS: 7789-42-6 | CdBr2

Cadmium bromide is used in process engraving, lithography and photography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-42-6
Molecular Formula CdBr2
MDL Number MFCD00015996
Synonym cadmium 2+ dibromide, cadmium 2+ ion dibromide, cadmium bromide, anhydrous 25g, cadmium bromide, anhydrous,

Specifications

Boiling Point 863°C
Melting Point 567°C
Color White to Yellow
Physical Form Powder
Quantity 100 g
Assay Percent Range 98.00%
Solubility Information Freely soluble in water,alcohol. Slightly soluble in acetone and ether.
Formula Weight 272.22
Odor Odorless
Sensitivity Hygroscopic
Appearance White to pale yellow powder
Density 5.192
Chemical Name or Material Cadmium bromide
Show More Show Less
Hazard Category H302+H312+H332-H350
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c
DOTInformation DOT Class: 6.1, Packaging Group: III
EINECSNumber 232-165-1
RTECSNumber EU9935000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.