Learn More
Cadmium bromide, ultra dry, 99.999% (metals basis)
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Specifications
Specifications
| Boiling Point | 863°C |
| Melting Point | 567°C |
| Color | White to Yellow |
| Physical Form | Beads |
| Quantity | 25 g |
| Odor | Odorless |
| Packaging | Ampule |
| Sensitivity | Hygroscopic |
| Density | 5.192 g/mL |
| UN Number | UN2570 |
| Show More |
Safety and Handling
| Hazard Category | H302+H312+H332-H350 |
| Hazard Statement | GHS H Statement H302-H312-H332 Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. |
| Precautionary Statement | P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS |
| EINECSNumber | 232-165-1 |
| RTECSNumber | EU9935000 |
| TSCA | Yes |
| storageNote1 |
|
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.