Learn More
Cadmium chloride, 98%, pure, anhydrous
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 960.0°C |
| Melting Point | 568.0°C |
| Quantity | 2.5 kg |
| Assay Percent Range | 97.5% min. (Argentometry) |
| Linear Formula | CdCl2 |
| Fieser | 04,485 |
| Merck Index | 15,1622 |
| Solubility Information | Solubility in water: 1400g/L (20°C). Other solubilities: soluble in acetone, slightly soluble in methanol and ethanol, practically insoluble in ether |
| Formula Weight | 183.31 |
| Percent Purity | 98% |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement May cause genetic defects. Causes damage to organs through prolonged or repeated exposure. Fatal if inhaled. May cause cancer. May damage fertility. May damage the unborn child. Toxic if swallowed |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Obtain special instructions before use. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF S |
| EINECSNumber | 233-296-7 |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.