Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium chloride, 99%, for analysis, anhydrous

Catalog No. AC219145000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC219145000 500 g Glass bottle
AC219141000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC219145000 Supplier Thermo Scientific Chemicals Supplier No. 219145000
Add to Cart

CAS: 10108-64-2 | CdCl2 | 183.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10108-64-2
Molecular Formula CdCl2
Molecular Weight (g/mol) 183.31
MDL Number MFCD00010916
InChI Key YKYOUMDCQGMQQO-UHFFFAOYSA-L
Synonym cadmium chloride, cadmium dichloride, caddy, vi-cad, kadmiumchlorid, caswell no. 135, ccris 114, hsdb 278, cadmium ii chloride
PubChem CID 24947
ChEBI CHEBI:35456
SMILES [Cl-].[Cl-].[Cd++]

Specifications

Boiling Point 960.0°C
Melting Point 568.0°C
Color White
Physical Form Powder
Quantity 500 g
Assay Percent Range 98.5% min. (On dry substance) (Argentometry)
Linear Formula CdCl2
Fieser 04,485
Merck Index 15, 1622
Solubility Information Solubility in water: 1400g/L (20°C). Other solubilities: soluble in acetone, slightly soluble in methanol and ethanol, practically insoluble in ether
IUPAC Name cadmium(2+) dichloride
Formula Weight 183.31
Percent Purity 99%
Grade Analytical
Packaging Glass bottle
Chemical Name or Material Cadmium chloride, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause genetic defects.
Causes damage to organs through prolonged or repeated exposure.
Fatal if inhaled.
May cause cancer.
May damage fertility.
May damage the unborn child.
Toxic if swallowed
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Obtain special instructions before use.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF S
EINECSNumber 233-296-7
RTECSNumber EV0175000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.