Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium Chloride, Solstice

Catalog No. 6000388 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-003-88 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-003-88 Supplier Solstice Supplier No. 20899100G

missing translation for 'validMainframeMsg'

Puriss. p.a., anhydrous,≥99.0%

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories: Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications. Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10108-64-2
Molecular Formula CdCl2
Molecular Weight (g/mol) 183.31
MDL Number MFCD00010916
InChI Key YKYOUMDCQGMQQO-UHFFFAOYSA-L
Synonym cadmium chloride, cadmium dichloride, caddy, vi-cad, kadmiumchlorid, caswell no. 135, ccris 114, hsdb 278, cadmium ii chloride
PubChem CID 24947
ChEBI CHEBI:35456
IUPAC Name dichlorocadmium
SMILES [Cl-].[Cl-].[Cd++]

Specifications

Physical Form Anhydrous
Quantity 100 g
UN Number 2570
Solubility Information Soluble in water 457g/L at 20°C
Formula Weight 183.32g/mol
Percent Purity ≥99%
Grade puriss. p.a.
Vapor Pressure 10mmHg (656°C)
Density Vapor Density: 6.3 (vs air)
Chemical Name or Material Cadmium Chloride
Hazard Statement P201-P260-P280-P304 + P340 + P310-P308 + P313
Precautionary Statement H301-H330-H340-H350-H360FD-H372-H410
EINECSNumber 233-296-7
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.