Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium chloride hydrate, Puratronic™, 99.998% (metals basis)

Catalog No. p-7048481
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1066109 10 g
AA1066118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1066109 Supplier Thermo Scientific Chemicals Supplier No. 01066109
Only null left
Add to Cart

CAS: 654054-66-7 | CdCl2 | 183.31 g/mol

Cadmium chloride hydrate is used in dyeing, printing, textiles and in metal finishing baths. It is also used as a reagent to make photographic paper and carbon paper. Further, it is utilized as an absorbing agent to absorb hydrogen sulfide as well as analytical reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 654054-66-7
Molecular Formula CdCl2
Molecular Weight (g/mol) 183.31
MDL Number MFCD00149627
InChI Key YKYOUMDCQGMQQO-UHFFFAOYSA-L
Synonym cadmium chloride, ultra dry, cdcl2.h2o, cadmium chloride hydrate, cadmium chloride 1-hydrate, cadmium chloride hydrate, puratronic, cadmium chloride monohydrate, puriss, cadmium chloride hydrate trace metals basis
PubChem CID 16211517
SMILES [Cl-].[Cl-].[Cd++]

Specifications

Physical Form Crystalline Powder
Quantity 10 g
UN Number UN2570
Merck Index 14,1617
Solubility Information Soluble in water and acetone. Slightly soluble in methanol. Insoluble in ether.
IUPAC Name cadmium(2+) dichloride
Formula Weight 183.32 (Anhydrous)
Percent Purity 99.998%
Grade Puratronic
Refractive Index 1.6513
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Cadmium chloride hydrate
Show More Show Less
Hazard Category H301-H330-H340-H350-H360-H372
Hazard Statement GHS H Statement
H300-H330-H340-H350-H360-H372
Fatal if swallowed.
Fatal if inhaled.
May cause genetic defects.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P284-P301+P310-P304+P340-P308+P313-P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CADMIUM COMPOUNDS
EINECSNumber 233-296-7
RTECSNumber EV0175000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.