Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium chloride, ultra dry, 99.996% (metals basis)

Catalog No. AA4285204
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
250 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4285204 2 g
AA4285209 10 g
AA4285218 50 g
AA4285230 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA4285204 Supplier Thermo Scientific Chemicals Supplier No. 04285204
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 10108-64-2 | CdCl2 | 183.31 g/mol

Cadmium chloride is used for the preparation of cadmium sulfide, used as Cadmium Yellow, a brilliant-yellow stable inorganic pigment. It can be used for the preparation of organocadmium compounds of the type R2Cd, where R is an aryl or a primary alkyl. It is also used for photocopying, dyeing and electroplating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10108-64-2
Molecular Formula CdCl2
Molecular Weight (g/mol) 183.31
MDL Number MFCD00010916
InChI Key YKYOUMDCQGMQQO-UHFFFAOYSA-L
Synonym cadmium chloride, cadmium dichloride, caddy, vi-cad, kadmiumchlorid, caswell no. 135, ccris 114, hsdb 278, cadmium ii chloride
PubChem CID 24947
ChEBI CHEBI:35456
IUPAC Name dichlorocadmium
SMILES [Cl-].[Cl-].[Cd++]

Specifications

Boiling Point 960°C
Melting Point 568°C
Color White
Physical Form Powder
Quantity 2 g
Odor Odorless
Sensitivity Hygroscopic
Appearance White Various forms (powder/flake/crystalline/beads
Assay Percent Range 99.996% (Metal basis)
Density 4.047 g/cm3 at 20°C
Solubility Information Soluble in water. Slightly soluble in alcohol.
Formula Weight 183.32
Chemical Name or Material Cadmium chloride
Show More Show Less
Hazard Category H301-H330-H335-H340-H350-H360FD-H372
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P284-P301+P310-P304+P340-P308+P313-P310-P330-P501c
DOTInformation DOT Class: 6.1, Packaging Group: III
EINECSNumber 233-296-7
RTECSNumber EV0175000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.