Learn More
Hach Company Cadmium column seal gasket - replacement rubber seal for Lachat cadmium column
Catalog No. NC1938309 Shop All Hach Company Products
Encompass
Replacement rubber gasket used to seal cadmium reduction columns for nitrate determination. Designed as a spare part for the cadmium reduction column assembly, it restores a leak-resistant seal and ensures proper flow through the column.
  • Provides a leak-resistant seal for cadmium reduction columns.
  • Made from resilient rubber to maintain compression and sealing properties.
  • Easy to install as a direct replacement gasket.
  • Sold individually for targeted maintenance and repair.
Catalog No. NC1938309 Supplier Hach Company Supplier No. 50252
Replacement rubber gasket used to seal cadmium reduction columns for nitrate determination. Designed as a spare part for the cadmium reduction column assembly, it restores a leak-resistant seal and ensures proper flow through the column.
  • Provides a leak-resistant seal for cadmium reduction columns.
  • Made from resilient rubber to maintain compression and sealing properties.
  • Easy to install as a direct replacement gasket.
  • Sold individually for targeted maintenance and repair.