Learn More
Hach Company Cadmium Granules, 40 Mesh, 10 g (Hach 50231)
Supplier:  Hach Company 50231
Encompass
Hach cadmium granules, 40 mesh, supplied in a 10 g package, provide cadmium metal in granular form for laboratory and analytical applications. The product is intended for reagent preparation and other processes requiring elemental cadmium. The item may be subject to hazardous-material shipping restrictions; follow applicable safety and shipping regulations.
  • Form: granules.
  • Mesh size: 40 mesh.
  • Net weight: 10 g.
  • Material: cadmium.
  • Intended use: laboratory and analytical reagent preparation.
  • Shipping: may be considered hazardous under some shipping conditions.
Catalog No. NC2221452