Learn More
Strem, An Ascensus Company CAS# 7440-43-9. 100g. Cadmium granules (99.999%). MFCD00010914
SDP
Catalog No. 5090115498
Encompass
CAS# 7440-43-9. 100g. Cadmium granules (99.999%). MFCD00010914. Molecular Weight: 112.41. Molecular Formula: Cd. Color/form: 1-5 mm Strem# 93-4851. http://www.strem.com/catalog/v/93-4851/
Catalog No. 50-901-15498 Supplier Strem, An Ascensus Company Supplier No. 934851100G
CAS# 7440-43-9. 100g. Cadmium granules (99.999%). MFCD00010914. Molecular Weight: 112.41. Molecular Formula: Cd. Color/form: 1-5 mm Strem# 93-4851. http://www.strem.com/catalog/v/93-4851/